Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ R3HCC1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA554710
Description
Immunogen sequence: ENDFVHRIQE ELDRFLLQKQ LSKVLLFPPL SSRLRYLIHR TAENFDLLSS FSVGEGWKRR TVICHQDIRV PSSDGLSGPC RAPASCPSRY HGPRPISNQG Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 87%.
R3HCC1 gene ontology annotations related to this gene include nucleic acid binding.
Specifications
R3HCC1 | |
Polyclonal | |
Unconjugated | |
R3HCC1 | |
1700020M16Rik; AA407796; R3H and coiled-coil domain-containing protein 1; R3H domain and coiled-coil containing 1; R3HCC1; RGD1311790 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
203069 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9Y3T6 | |
R3HCC1 | |
Recombinant protein corresponding to Human R3HCC1. Recombinant protein control fragment (Product #RP-93754). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction