Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R3HDM1 Antibody, Alexa Fluor™ 750, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP241362AF750
Description
R3HDM1 Polyclonal antibody specifically detects R3HDM1 in Human, Mouse samples. It is validated for Western BlotSpecifications
R3HDM1 | |
Polyclonal | |
Western Blot | |
R3H domain containing 1, R3H domain-containing protein 1 | |
The epitope is recognized to a region between residue 500 to 550 of human R3H domain-containing protein 1 using the numbering given in entry Q15032 (GeneID 23518). | |
0.1 mL | |
Primary | |
Human, Mouse | |
Purified |
Western Blot | |
Alexa Fluor 750 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
23518 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction