Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ R3HDM2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA558346
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA558346 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA558346 Supplier Invitrogen™ Supplier No. PA558346
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: YSGVSPSGPG VVVMQLNVPN GPQPPQNPSM VQWSHCKYYS MDQRGQKPGD LYSPDSSPQA NTQMSSSPVT SPTQSP Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 89%.

The R3HDM2 (R3H Domain Containing 2) gene encodes a protein that contains an R3H domain, which is known to bind single-stranded nucleic acids. Located on chromosome 12, R3HDM2 has been implicated in various cellular processes, including RNA binding and regulation of gene expression. The R3H domain is characterized by the ability to recognize specific sequences or structures in RNA or DNA, suggesting that R3HDM2 may play a role in post-transcriptional regulation, mRNA processing, or DNA repair. The precise biological functions and mechanisms of R3HDM2 are still under investigation, but its potential involvement in these fundamental processes indicates its importance in maintaining cellular homeostasis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen R3HDM2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene R3HDM2
Gene Accession No. Q9Y2K5
Gene Alias CAG6; KIAA1002; PR01365; R3H domain containing 2; R3H domain-containing protein 2; R3HDM2
Gene Symbols R3HDM2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human R3HDM2. Recombinant protein control fragment (Product #RP-97852).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22864
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.