Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R718 Mouse Anti-Human IL-13 Receptor α2 (CD213a2), BD Horizon™
Mouse Monoclonal Antibody
$188.00 - $468.00
Specifications
Antigen | IL-13 Receptor α2 (CD213a2) |
---|---|
Clone | 47 (also known as 47; mAb47; 47 mAb) |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | R718 |
Description
IL-13 Receptor α2 (CD213a2) Monoclonal antibody specifically detects IL-13 Receptor α2 (CD213a2) in Human samples. It is validated for Flow cytometrySpecifications
IL-13 Receptor α2 (CD213a2) | |
Flow Cytometry | |
R718 | |
Mouse | |
Human | |
CD213A2; IL-13 receptor subunit alpha-2; IL-13R; IL-13Rα2; cancer/testis antigen 19 | |
Human rhIL13Ra2hFc | |
Affinity Chromatography |
47 (also known as 47; mAb47; 47 mAb) | |
Monoclonal | |
Liquid | |
RUO | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
3598 | |
IgG1 κ | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title