Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R718 Rat Anti-Mouse Granzyme C, BD Horizon™
Rat Monoclonal Antibody
$122.00 - $306.00
Specifications
Antigen | Granzyme C |
---|---|
Clone | SFC1D8.rMAb |
Concentration | 0.2 mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Description
Granzyme C Monoclonal antibody specifically detects Granzyme C in Mouse samples. It is validated for Intracellular staining (flow cytometry)Specifications
Granzyme C | |
0.2 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
14940 | |
IgG1 κ | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
SFC1D8.rMAb | |
Flow Cytometry | |
R718 | |
Rat | |
Mouse | |
Gzmc; cytotoxic cell protease 2; CCP2; Ctla5; Ctla-5 | |
Purified Recombinant Mouse Granzyme C Protein | |
Affinity Chromatography |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title