Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R718 Rat Anti-TOX, BD Horizon™
Rat Monoclonal Antibody
Supplier: BD Biosciences 569822
Description
TOX Monoclonal antibody specifically detects TOX in Human|Mouse samples. It is validated for Intracellular staining (flow cytometry)
Specifications
TOX | |
Monoclonal | |
0.2 mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Rat | |
Affinity Chromatography | |
RUO | |
Human, Mouse | |
Liquid |
Flow Cytometry | |
NAN448B | |
R718 | |
TOX; TOX 1; TOX1 | |
Recombinant Human TOX Protein | |
0.1 mg | |
9760, 252838 | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction