Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ R9BP Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA562047
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562047 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562047 Supplier Invitrogen™ Supplier No. PA562047
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LDGLNKTTAC YHHLVLTVGG SADSQDLRQE LQKTRQKAQE LAVSTCARLT AVLRDRGLAA DERAEFERLW VAFSGCLDLL EAD Highest antigen sequence identity to the following orthologs: Mouse - 87%, Rat - 84%.

The protein encoded by this gene functions as a regulator of G protein-coupled receptor signaling in phototransduction. Studies in bovine and mouse show that this gene is expressed only in the retina, and is localized in the rod outer segment membranes. This protein is associated with a heterotetrameric complex, specifically interacting with the regulator of G-protein signaling 9, and appears to function as the membrane anchor for the other largely soluble interacting partners. Mutations in this gene are associated with prolonged electroretinal response suppression (PERRS), also known as bradyopsia.
TRUSTED_SUSTAINABILITY

Specifications

Antigen R9BP
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RGS9BP
Gene Accession No. Q6ZS82
Gene Alias membrane anchor R9AP; PERRS; R9AP; regulator of G protein signaling 9 binding protein; regulator of G-protein signaling 9 binding protein; regulator of G-protein signaling 9-binding protein; regulator of G-protein signalling 9 binding protein; RGD1561747; RGS9; RGS9 anchor protein; RGS9-1 anchor; Rgs9-1bp; RGS9-anchoring protein; RGS9BP
Gene Symbols RGS9BP
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human R9BP. Recombinant protein control fragment (Product #RP-99979).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 388531
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.