Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ RA 839
GREENER_CHOICESDP

Catalog No. 570750 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
57-075-0 50 mg
57-071-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 57-075-0 Supplier Tocris Bioscience™ Supplier No. 5707/50
Add to Cart
Add to Cart
This item is not returnable. View return policy

Nrf2 activator; inhibits Nrf2/Keap1 interaction

RA 839 is an Nrf2 activator; inhibits Nrf2/Keap1 interaction (Kd = 6 μM). Suppresses LPS-induced iNOS and nitric oxide expression in macrophages. Anti-inflammatory.

Specifications

Chemical Name or Material (3S)-1-[4-[[(2,3,5,6-Tetramethylphenyl)sulfonyl]amino]-1-naphthalenyl]-3-pyrrolidinecarboxylic acid
CAS 1832713-02-6
Quantity 50 mg
Target Nrf2 Modulators
Molecular Formula C25H28N2O4S
Formula Weight 452.57
Purity >98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.