Learn More
CD45 Mouse anti-Rat, Brilliant Violet 711, Clone: OX-1, BD Optibuild™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 740725
Description
The OX-1 antibody recognizes all molecular forms of CD45 (Leukocyte Common Antigen) on all hematopoietic cells except erythrocytes. CD45 is a member of the Protein Tyrosine Phosphatase (PTP) family; its intracellular (COOH-terminal) region contains two PTP catalytic domains while the extracellular region is highly variable due to alternative splicing of exons 4, 5, and 6 (designated A, B, and C, respectively), and differing levels of glycosylation. The CD45 isoforms detected in rat are cell type-, maturation-, and activation state-specific. The CD45 isoforms play complex roles in T-cell and B-cell antigen receptor signal transduction.
The antibody was conjugated to BD Horizon™ BV711 which is part of the BD Horizon Brilliant™ Violet family of dyes. This dye is a tandem fluorochrome of BD Horizon BV421 with an Ex Max of 405-nm and an acceptor dye with an Em Max at 711-nm. BD Horizon BV711 can be excited by the violet laser and detected in a filter used to detect Cy™5.5 / Alexa Fluor™ 700-like dyes (eg, 712/20-nm filter). Due to the excitation and emission characteristics of the acceptor dye, there may be moderate spillover into the Alexa Fluor™ 700 and PerCP-Cy5.5 detectors. However, the spillover can be corrected through compensation as with any other dye combination.

Specifications
CD45 | |
Monoclonal | |
0.2mg/mL | |
Ptprc; Lca; Leucocyte common antigen; RT7; T200 | |
Mouse | |
Affinity Purified | |
RUO | |
Rat | |
IgG1 κ |
Flow Cytometry | |
OX-1 | |
Brilliant Violet 711 | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Leukocyte Common Antigen-enriched Glycoprotein Fraction from Wistar Rat Thymocytes | |
50 μg | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only