Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RAB-14 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501985023
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-5023 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-5023 Supplier Bioss Supplier No. BS2933R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Localized to biosynthetic and endosomal compartments, Rab14 is involved in the biosynthetic/recycling pathway between the Golgi and endosomal compartments.

Specifications

Antigen RAB-14
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene RAB14
Gene Accession No. P61106
Gene Alias 0610030G24Rik; 2810475J17Rik; A830021G03Rik; AI314285; AI649155; bA165P4.3 (member RAS oncogene family); BOS_9407; cb731; D030017L14Rik; F protein-binding protein 1; FBP; fc26g11; fj47f07; fj68a01; GTPase Rab14; Rab14; RAB-14; RAB14, member RAS oncogene family; Ras-related protein Rab-14; small GTP binding protein RAB14; wu:fc26g11; wu:fj47f07; wu:fj68a01
Gene Symbols RAB14
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human RAB-14.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51552
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.