Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RAB10 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5110252
Description
Immunogen sequence: ENISKWLRNI DEHANEDVER MLLGNKCDMD DKRVVPKGKG EQIAREHGIR FFETSAKANI NIEKAFLTLA EDILRKTPVK EPNSENVDIS SGGGVTGWKS K.
RAB10 belongs to the RAS superfamily of small GTPases. RAB proteins localize to exocytic and endocytic compartments and regulate intracellular vesicle trafficking.
Specifications
| RAB10 | |
| Polyclonal | |
| Unconjugated | |
| RAB10 | |
| Ac1075; AW107754; GTP-binding protein RAB10; Rab10; RAB10, member RAS oncogene family; ras-related GTP-binding protein; ras-related protein rab10; ras-related protein Rab-10 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 10890 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 1.96 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P61026 | |
| RAB10 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 98-198 of human RAB10 (NP_0572153). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction