Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Rab11 FIB3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5116482
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5116482 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5116482 Supplier Invitrogen™ Supplier No. PA5116482
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive controls: MCF-7 cell lysates, mouse liver tissue, mouse kindey tissue, MCF-7.

Proteins of the large Rab GTPase family have regulatory roles in the formation, targeting, and fusion of intracellular transport vesicles. RAB11FIP3 is one of many proteins that interact with and regulate Rab GTPases (Hales et al., 2001 ).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Rab11 FIB3
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4
Gene RAB11FIP3
Gene Accession No. O75154, Q8CHD8
Gene Alias ARFO1; Arfophilin-1; Cart1; cytoplasmic adaptor for RAR and TR; D030060O14Rik; EF hands-containing Rab-interacting protein; eferin; FIP3-Rab11; Kiaa0665; LRRGT00043; mKIAA0665; MU-MB-17.148; PAC196A12.1; RAB11 family interacting protein 3; RAB11 family interacting protein 3 (class II); rab11 family-interacting protein 3; rab11-family interacting protein 3; Rab11fip3; Rab11-FIP3; RGD1308952
Gene Symbols RAB11FIP3
Host Species Rabbit
Immunogen Synthetic peptide within mouse FIP3 aa 543-592.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 215445, 9727
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.