Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Rab11A Antibody (STo 7C10), Alexa Fluor™ 594, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP250260AF594
Description
Rab11A Monoclonal specifically detects Rab11A in Human samples. It is validated for Western Blot, ELISA.Specifications
Rab11A | |
Monoclonal | |
Alexa Fluor 594 | |
50 mM sodium borate with 0.05% sodium azide | |
RAB11A | |
Rab11a peptide KQMSDRRENDM | |
0.1 mL | |
Primary | |
Human | |
IgG1 |
Western Blot, ELISA | |
STo 7C10 | |
Western Blot, ELISA | |
MGC1490, RAB 11A, member oncogene family, Rab-11, RAB11, RAB11A, member RAS oncogene family, YL8ras-related protein Rab-11A | |
Mouse | |
Protein A purified | |
Cancer, Tumor Suppressors | |
8766.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction