Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RAB11B Antibody, Alexa Fluor™ 750, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP318858AF750
Description
RAB11B Polyclonal antibody specifically detects RAB11B in Human samples. It is validated for Western Blot, ImmunofluorescenceSpecifications
RAB11B | |
Polyclonal | |
Western Blot, Immunocytochemistry/Immunofluorescence | |
GTP-binding protein YPT3, H-YPT3, MGC133246, RAB11B, member of RAS oncogene family, RAB11B, member RAS oncogene family, ras-related protein Rab-11B, YPT3 | |
Produced in rabbits immunized with purified, recombinant Human RAB11B (Accesion # ABQ59034.1, Gly2-Cys215) | |
0.1 mL | |
Cancer, GPCR, Signal Transduction | |
9230 | |
Store at 4C in the dark. | |
IgG |
Western Blot, Immunofluorescence | |
Alexa Fluor 750 | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction