Learn More
Invitrogen™ RAB11B Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA1775
Description
PA1-775 detects Rab 11 from human and non-human primate samples. This antibody detects both A and B isoforms. PA1-775 has been successfully used in Western blot procedures. By Western blot, this antibody detects an ~22 kDa protein representing Rab 11 in CV1, PC3, and HeLa cell extract. Another less prominent, unknown band is also detected by this antibody at ~35 kDa. The PA1-775 immunizing peptide corresponds to amino acid residues 1-17 from human Rab 11. This sequence is completely conserved in human, mouse and rat and 94% conserved in C. elegans and amphibians. This peptide (Cat. # PEP-195) is available for use in neutralization and control experiments.
Rab proteins are low-molecular-weight GTP-binding proteins that form the largest branch of the Ras superfamily of GTPases. Located on the cytoplasmic face of organelles and vesicles, rab proteins are involved in intracellular membrane fusion reactions. Three membrane proteins, synaptosomal associated protein of 25 kDa (SNAP-25), synaptobrevin, and syntaxin, form the core of a ubiquitous membrane fusion machine that interacts with the soluble proteins N-ethylmaleimide-sensitive factor (NSF) and a-SNAP. Rab proteins, in coordination with the core fusion machinery and Munc-18, help to mediate vesicle docking and fusion. There exist over 40 Rab proteins that have been described in mammals. Rab 11 is a known marker for protein trafficking, sorting, and recycling in the endosomal pathway.
Specifications
RAB11B | |
Polyclonal | |
Unconjugated | |
Rab11b | |
A730055L17Rik; GTP-binding protein YPT3; H-YPT3; RAB11B; RAB11B, member of RAS oncogene family; RAB11B, member RAS oncogene family; RAS oncogene family protein; ras-related protein Rab-11B; YPT3 | |
Rabbit | |
Affinity chromatography | |
RUO | |
611619, 9230 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 1 mg/mL BSA and 0.05% sodium azide | |
Q15907 | |
Rab11b | |
Synthetic peptide corresponding to residues M(1) G T R D D E Y D Y L F K V V L I C(17) of human Rab11. | |
200 μg | |
Primary | |
Human, Canine, Monkey | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.