Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RAB11FIP1 Mouse anti-Human, Mouse, Clone:3A12H9D2, Abnova™

Catalog No. 89333068 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-333-068 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-333-068 Supplier Abnova Corporation Supplier No. MAB16736
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against recombinant human RAB11FIP1.

Proteins of the large Rab GTPase family (see RAB1A; MIM 179508) have regulatory roles in the formation, targeting, and fusion of intracellular transport vesicles. RAB11FIP1 is one of many proteins that interact with and regulate Rab GTPases (Hales et al., 2001 [PubMed 11495908]).[supplied by OMIM]

Specifications

Antigen RAB11FIP1
Applications ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 3A12H9D2
Conjugate Unconjugated
Description RAB11 family interacting protein 1 (class I)
Dilution ELISA (1:10000)
Western Blot (1:500-1:2000)
Immunohistochemistry (1:200-1:1000)
Immunocytochemistry
Flow Cytometry (1:200-1:400)
The optimal working dilution should be determined by the end user.
Formulation In PBS (0.05% sodium azide)
Gene RAB11FIP1
Gene Alias DKFZp686E2214, FLJ22524, FLJ22622, MGC78448, NOEL1A, RCP, rab11-FIP1
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acid 130-271 of human RAB11FIP1 from E. coli.
Quantity 100 μg
Gene ID (Entrez) 80223
Target Species Human, Mouse
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.