Learn More
Invitrogen™ RAB11FIP1 Polyclonal Antibody

Description
Antibody detects endogenous levels of total RAB11FIP1.
Specifications
Specifications
| Antigen | RAB11FIP1 |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide |
| Gene | RAB11FIP1 |
| Gene Accession No. | Q6WKZ4, Q9D620 |
| Gene Alias | 2010200K21Rik; 4833414G05Rik; CBBM; CBP; hCG_41347; NOEL1A; Rab coupling protein; Rab effector protein; RAB11 coupling protein; RAB11 family interacting protein 1; RAB11 family interacting protein 1 (class I); rab11 family-interacting protein 1; RAB11FIP1; Rab11-FIP1; rab11fip1 {ECO:0000250; rab-coupling protein; Rab-interacting recycling protein; Rcp; RGD1562965; UniProtKB:Q6WKZ4} |
| Gene Symbols | RAB11FIP1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.