Learn More
Invitrogen™ RAB11FIP1 Polyclonal Antibody

Description
Additional Information: This antibody is expected to recognise isoform 1 (NP_079427.3) and isoform 3 (NP_001002814.1). This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:64,000.
Specifications
Specifications
| Antigen | RAB11FIP1 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 |
| Gene | RAB11FIP1 |
| Gene Accession No. | Q6WKZ4 |
| Gene Alias | 2010200K21Rik; 4833414G05Rik; CBBM; CBP; hCG_41347; NOEL1A; Rab coupling protein; Rab effector protein; RAB11 coupling protein; RAB11 family interacting protein 1; RAB11 family interacting protein 1 (class I); rab11 family-interacting protein 1; RAB11FIP1; Rab11-FIP1; rab11fip1 {ECO:0000250; rab-coupling protein; Rab-interacting recycling protein; Rcp; RGD1562965; UniProtKB:Q6WKZ4} |
| Gene Symbols | RAB11FIP1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.