Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RAB11FIP2 Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA550102

Catalog No. PA550102


Only null left
Add to Cart

Description

Description

The antibody detects endogenous levels of total RAB11FIP2 protein.

RAB11FIP2 is an adapter protein that plays a role in the secretory pathway. It is thought to be important for endosome recycling and receptor-mediated endocytosis. In endosome recycling, RAB11-FIP2 regulates vesicle transport from the endosomal recycling compartment (ERC) to the plasma membrane.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

RAB11FIP2
Polyclonal
Unconjugated
RAB11FIP2
4930470G04Rik; A830046J09Rik; AW558126; KIAA0941; nRip11; RAB11 family interacting protein 2; RAB11 family interacting protein 2 (class I); rab11 family-interacting protein 2; rab11-family interacting protein 2; Rab11fip2; Rab11-FIP2; RAB11-FIP2 long isoform; RGD1308538
Rabbit
Antigen affinity chromatography
RUO
22841, 74998
-20°C
Liquid
Western Blot
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
G3XA57, Q7L804
RAB11FIP2
Synthesized peptide derived from internal of human RAB11FIP2.
100 μL
Primary
Human, Mouse
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.