Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RAB11FIP2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5106713
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5106713 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5106713 Supplier Invitrogen™ Supplier No. PA5106713
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total RAB11FIP2.

RAB11FIP2 is an adapter protein that plays a role in the secretory pathway. It is thought to be important for endosome recycling and receptor-mediated endocytosis. In endosome recycling, RAB11-FIP2 regulates vesicle transport from the endosomal recycling compartment (ERC) to the plasma membrane.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RAB11FIP2
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene RAB11FIP2
Gene Accession No. G3XA57, Q7L804
Gene Alias 4930470G04Rik; A830046J09Rik; AW558126; KIAA0941; nRip11; RAB11 family interacting protein 2; RAB11 family interacting protein 2 (class I); rab11 family-interacting protein 2; rab11-family interacting protein 2; Rab11fip2; Rab11-FIP2; RAB11-FIP2 long isoform; RGD1308538
Gene Symbols RAB11FIP2
Host Species Rabbit
Immunogen A synthesized peptide derived from human RAB11FIP2(Accession Q7L804), corresponding to amino acid residues F339-P389.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22841, 308003, 74998
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.