Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RAB11FIP3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA556335
Description
Immunogen sequence: QVKDLTKYLD PSGLGVISFE DFYQGITAIR NGDPDGQCYG GVASAQDEEP LACPDEFDDF VTYEANEVTD SAYMGSESTY SECETF Highest antigen sequence identity to the following orthologs: Mouse - 87%, Rat - 27%.
Proteins of the large Rab GTPase family have regulatory roles in the formation, targeting, and fusion of intracellular transport vesicles. RAB11FIP3 is one of many proteins that interact with and regulate Rab GTPases (Hales et al., 2001 ).
Specifications
RAB11FIP3 | |
Polyclonal | |
Unconjugated | |
RAB11FIP3 | |
ARFO1; Arfophilin-1; Cart1; cytoplasmic adaptor for RAR and TR; D030060O14Rik; EF hands-containing Rab-interacting protein; eferin; FIP3-Rab11; Kiaa0665; LRRGT00043; mKIAA0665; MU-MB-17.148; PAC196A12.1; RAB11 family interacting protein 3; RAB11 family interacting protein 3 (class II); rab11 family-interacting protein 3; rab11-family interacting protein 3; Rab11fip3; Rab11-FIP3; RGD1308952 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
9727 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O75154 | |
RAB11FIP3 | |
Recombinant protein corresponding to Human RAB11FIP3. Recombinant protein control fragment (Product #RP-95105). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction