Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RAB11FIP3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 258431AP150UL

Description
Proteins of the large Rab GTPase family have regulatory roles in the formation, targeting, and fusion of intracellular transport vesicles. RAB11FIP3 is one of many proteins that interact with and regulate Rab GTPases (Hales et al., 2001 ).Specifications
RAB11FIP3 | |
Polyclonal | |
Unconjugated | |
RAB11FIP3 | |
ARFO1, Arfophilin 1, Eferin, FIP3 Rab11, KIAA0665, MU MB 17.148, Rab11 FIP3, RAB11FIP3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
215445, 303002, 9727 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.26 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O75154, Q8CHD8 | |
RAB11FIP3 | |
RAB11FIP3 Fusion Protein Ag22937 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction