Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RAB11FIP4 Antibody (OTI13B5), DyLight 755, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273764IR
Description
RAB11FIP4 Monoclonal specifically detects RAB11FIP4 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
RAB11FIP4 | |
Monoclonal | |
DyLight 755 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
RAB11FIP4 | |
Recombinant protein fragment corresponding to amino acids 91-177 of human RAB11FIP4 (NP_116321) produced in E.coli. | |
0.1 mL | |
84440.0 | |
Store at 4°C in the dark. |
Western Blot, Immunohistochemistry | |
OTI13B5 | |
Western Blot, Immunohistochemistry | |
ARFO2, Arfophilin-2, FIP4-Rab11, FLJ00131, KIAA1821arfophilin-2, MGC11316, MGC126566, RAB11 family interacting protein 4 (class II), rab11 family-interacting protein 4, Rab11-FIP4 | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction