Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RAB11FIP4 Monoclonal Antibody (OTI13B5)
GREENER_CHOICE

Catalog No. PIMA526978
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA526978 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA526978 Supplier Invitrogen™ Supplier No. MA526978
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

RAB11FIP4 Monoclonal Antibody for Western Blot, IHC (P)

Proteins of the large Rab GTPase family have regulatory roles in the formation, targeting, and fusion of intracellular transport vesicles. RAB11FIP4 is one of many proteins that interact with and regulate Rab GTPases.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RAB11FIP4
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI13B5
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene RAB11FIP4
Gene Accession No. Q86YS3
Gene Alias A730072L08Rik; ARFO2; Arfophilin-2; FIP4-Rab11; Kiaa1821; mKIAA1821; mRab11-FIP4A; RAB11 family interacting protein 4; RAB11 family interacting protein 4 (class II); rab11 family-interacting protein 4; Rab11fip4; RAB11-FIP4
Gene Symbols RAB11FIP4
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 91-177 of RAB11FIP4 produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84440
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.