Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RAB11FIP4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA554465
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA554465 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA554465 Supplier Invitrogen™ Supplier No. PA554465
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VLSVESAGTL PCAPEIPDCV EQGSEVTGPT FADGELIPRE PGFFPEDEEE AMTLAPPEGP QELYTDSPME STQSLEGSVG SPAEKDG Highest antigen sequence identity to the following orthologs: Mouse - 73%, Rat - 76%.

Proteins of the large Rab GTPase family have regulatory roles in the formation, targeting, and fusion of intracellular transport vesicles. RAB11FIP4 is one of many proteins that interact with and regulate Rab GTPases.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RAB11FIP4
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RAB11FIP4
Gene Accession No. Q86YS3
Gene Alias A730072L08Rik; ARFO2; Arfophilin-2; FIP4-Rab11; Kiaa1821; mKIAA1821; mRab11-FIP4A; RAB11 family interacting protein 4; RAB11 family interacting protein 4 (class II); rab11 family-interacting protein 4; Rab11fip4; RAB11-FIP4
Gene Symbols RAB11FIP4
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human RAB11FIP4. Recombinant protein control fragment (Product #RP-93254).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84440
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.