Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RAB11FIP5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA557642
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA557642 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA557642 Supplier Invitrogen™ Supplier No. PA557642
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SPHPVKPLSA APVEGSPDRK QSRSSLSIAL SSGLEKLKTV TSGSIQPVTQ APQAGQMVDT KRLKDSAVLD QSAKYYHLTH DELISLLLQR ERELSQRDEH VQEL Highest antigen sequence identity to the following orthologs: Mouse - 85%, Rat - 87%.

Rab effector involved in protein trafficking from apical recycling endosomes to the apical plasma membrane. Involved in insulin granule exocytosis. May regulate V-ATPase intracellular transport in response to extracellular acidosis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RAB11FIP5
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RAB11FIP5
Gene Accession No. Q9BXF6
Gene Alias 9130206P09Rik; C75969; D6Ertd32e; GAF1; Gaf-1; Gamma-SNAP-associated factor 1; KIAA0857; Phosphoprotein pp75; pp75; RAB11 family interacting protein 5; RAB11 family interacting protein 5 (class I); rab11 family-interacting protein 5; Rab11fip5; Rab11-FIP5; rab11-interacting protein Rip11; Rip11
Gene Symbols RAB11FIP5
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human RAB11FIP5. Recombinant protein control fragment (Product #RP-96081).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26056
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.