Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RAB11FIP5 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
Antigen | RAB11FIP5 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
RAB11FIP5 Polyclonal specifically detects RAB11FIP5 in Human samples. It is validated for Western Blot.Specifications
RAB11FIP5 | |
Polyclonal | |
Rabbit | |
NP_056285 | |
26056 | |
Synthetic peptide directed towards the N terminal of human RAB11FIP5The immunogen for this antibody is RAB11FIP5. Peptide sequence MALVRGAEPAAGPSRWLPTHVQVTVLRARGLRGKSSGAGSTSDAYTVIQV. | |
Primary |
Western Blot | |
Unconjugated | |
RUO | |
DKFZp434H018, Gaf-1, GAF1rab11-FIP5, Gamma-SNAP-associated factor 1, KIAA0857gaf-1, Phosphoprotein pp75, pp75, RAB11 family interacting protein 5 (class I), Rab11-FIP5, Rab11-interacting protein Rip11, RAB11RIP5, RIP11rab11 family-interacting protein 5 | |
RAB11FIP5 | |
IgG | |
70 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title