Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RAB12 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559270
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559270 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559270 Supplier Invitrogen™ Supplier No. PA559270
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ITKKETFDDL PKWMKMIDKY ASEDAELLLV GNKLDCETDR EITRQQGEKF AQQITGMRFC EASA Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 98%.

The small GTPases Rab are key regulators of intracellular membrane trafficking, from the formation of transport vesicles to their fusion with membranes. Rabs cycle between an inactive GDP-bound form and an active GTP-bound form that is able to recruit to membranes different set of downstream effectors directly responsible for vesicle formation, movement, tethering and fusion. That Rab may play a role in protein transport from recycling endosomes to lysosomes regulating, for instance, the degradation of the transferrin receptor. Involved in autophagy.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RAB12
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RAB12
Gene Accession No. Q6IQ22
Gene Alias 2900054P15Rik; AW555448; C77700; putative Ras-related protein Rab-12; Rab12; RAB12, member RAS oncogene family; rab-13; ras-related protein Rab-12
Gene Symbols RAB12
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human RAB12. Recombinant protein control fragment (Product #RP-98294).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 201475
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.