Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RAB13 Mouse anti-Human, Clone:8E8E2, Abnova™

Catalog No. 89333062 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-333-062 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-333-062 Supplier Abnova Corporation Supplier No. MAB16744
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against recombinant human RAB13.

Specifications

Antigen RAB13
Applications ELISA, Flow Cytometry, Immunocytochemistry, Western Blot
Classification Monoclonal
Clone 8E8E2
Conjugate Unconjugated
Description RAB13, member RAS oncogene family
Dilution ELISA (1:10000)
Western Blot (1:500-1:2000)
Immunohistochemistry
Immunocytochemistry (1:200-1:1000)
Flow Cytometry (1:200-1:400)
The optimal working dilution should be determined by the end user.
Formulation In PBS (0.05% sodium azide)
Gene RAB13
Gene Alias GIG4
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acid 66-200 of human RAB13 from E. coli.
Quantity 100 μg
Gene ID (Entrez) 5872
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.