Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RAB13 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA513621
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA513621 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA513621 Supplier Invitrogen™ Supplier No. PA513621
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody is predicted to react with bovine based on sequence homology.

RAB13 can participate in polarized transport, in the assembly and/or the activity of tight junctions.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RAB13
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 2 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene RAB13
Gene Accession No. P51153, Q9DD03
Gene Alias 0610007N03Rik; B230212B15Rik; Cell growth-inhibiting gene 4 protein; GIG4; growth-inhibiting gene 4 protein; Rab13; RAB13, member RAS oncogene family; RAS-associated protein RAB13; ras-related protein Rab-13
Gene Symbols RAB13
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 109-137 amino acids from the central region of human RAB13.
Purification Method Ammonium sulfate precipitation, Size-exclusion - Dialysis
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5872, 68328
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.