Learn More
Invitrogen™ RAB13 Polyclonal Antibody

Description
Immunogen sequence: MAKAYDHLFK LLLIGDSGVG KTCLIIRFAE DNFNNTYIST IGIDFKIRTV DIEGKKIKLQ VWDTAGQERF KTITTAYYRG AMGIILVYDI TDEKSFENIQ NWMKSIKENA SAGVERLLLG NKCDMEAKRK VQKEQADKLA REHGIRFFET SAKSSMNVDE AFSSLARDIL LKSGGRRSGN GNKPPSTDLK TCDKKNTNKC SLG; Positive Samples: HeLa, 293T, LO2; Cellular Location: Cell junction, Cell membrane, Cell projection, Cytoplasmic side, Cytoplasmic vesicle, Cytoplasmic vesicle membrane, Golgi apparatus, Lipid-anchor, Recycling endosome membrane, lamellipodium, secretory vesicle membrane, tight junction, trans-Golgi network membrane.
Specifications
Specifications
| Antigen | RAB13 |
| Applications | ELISA, Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1.02 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 |
| Gene | RAB13 |
| Gene Accession No. | P51153 |
| Gene Alias | 0610007N03Rik; B230212B15Rik; Cell growth-inhibiting gene 4 protein; GIG4; growth-inhibiting gene 4 protein; Rab13; RAB13, member RAS oncogene family; RAS-associated protein RAB13; ras-related protein Rab-13 |
| Gene Symbols | RAB13 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.