Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RAB14 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5100175
Description
Antibody detects endogenous levels of total RAB14.
Localized to biosynthetic and endosomal compartments, Rab14 is involved in the biosynthetic/recycling pathway between the Golgi and endosomal compartments.
Specifications
RAB14 | |
Polyclonal | |
Unconjugated | |
RAB14 | |
0610030G24Rik; 2810475J17Rik; A830021G03Rik; AI314285; AI649155; bA165P4.3 (member RAS oncogene family); cb731; D030017L14Rik; F protein-binding protein 1; FBP; fc26g11; fj47f07; fj68a01; GTPase Rab14; Rab14; RAB-14; RAB14, member RAS oncogene family; ras-related protein Rab-14; small GTP binding protein RAB14; wu:fc26g11; wu:fj47f07; wu:fj68a01 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
51552, 68365, 94197 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P61106, P61107, Q91V41 | |
RAB14 | |
A synthesized peptide derived from human RAB14(Accession P61106), corresponding to amino acid residues S180-C215. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction