Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RAB14 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5110253
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5110253 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5110253 Supplier Invitrogen™ Supplier No. PA5110253
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DCPHTIGVEF GTRIIEVSGQ KIKLQIWDTA GQERFRAVTR SYYRGAAGAL MVYDITRRST YNHLSSWLTD ARNLTNPNTV IILIGNKADL EAQRDVTYEE AK.

Localized to biosynthetic and endosomal compartments, Rab14 is involved in the biosynthetic/recycling pathway between the Golgi and endosomal compartments.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RAB14
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.49 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3
Gene RAB14
Gene Accession No. P61106, Q91V41
Gene Alias 0610030G24Rik; 2810475J17Rik; A830021G03Rik; AI314285; AI649155; bA165P4.3 (member RAS oncogene family); cb731; D030017L14Rik; F protein-binding protein 1; FBP; fc26g11; fj47f07; fj68a01; GTPase Rab14; Rab14; RAB-14; RAB14, member RAS oncogene family; ras-related protein Rab-14; small GTP binding protein RAB14; wu:fc26g11; wu:fj47f07; wu:fj68a01
Gene Symbols RAB14
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 39-140 of human RAB14 (NP_0574062).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51552, 68365
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.