Learn More
Invitrogen™ RAB15 Polyclonal Antibody

Description
Predicted to react with bovine, mouse and rat based on sequence homology.
Specifications
Specifications
| Antigen | RAB15 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | RAB15 |
| Gene Accession No. | P59190 |
| Gene Alias | 2310012G06Rik; AI840042; Rab15; RAB15, member RAS oncogene family; RAB15, member RAS onocogene family; ras-related protein Rab-15; Ras-related protein Rab-15 isoform AN1; Ras-related protein Rab-15 isoform AN2; Ras-related protein Rab-15 isoform AN3 |
| Gene Symbols | RAB15 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.