Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RAB15 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA568300
Description
Predicted to react with Human and Rat samples.
May act in concert with RAB3A in regulating aspects of synaptic vesicle membrane flow within the nerve terminal.
Specifications
RAB15 | |
Polyclonal | |
Unconjugated | |
RAB15 | |
2310012G06Rik; AI840042; Rab15; RAB15, member RAS oncogene family; RAB15, member RAS onocogene family; ras-related protein Rab-15; Ras-related protein Rab-15 isoform AN1; Ras-related protein Rab-15 isoform AN2; Ras-related protein Rab-15 isoform AN3 | |
Rabbit | |
affinity chromatography | |
RUO | |
104886, 299156, 376267 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P35289, P59190, Q8K386 | |
RAB15 | |
Synthesized peptide derived from human RAB15. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction