Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RAB15 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5104065
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5104065 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5104065 Supplier Invitrogen™ Supplier No. PA5104065
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total RAB15.

May act in concert with RAB3A in regulating aspects of synaptic vesicle membrane flow within the nerve terminal.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RAB15
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene RAB15
Gene Accession No. P35289, P59190, Q8K386
Gene Alias 2310012G06Rik; AI840042; Rab15; RAB15, member RAS oncogene family; RAB15, member RAS onocogene family; ras-related protein Rab-15; Ras-related protein Rab-15 isoform AN1; Ras-related protein Rab-15 isoform AN2; Ras-related protein Rab-15 isoform AN3
Gene Symbols RAB15
Host Species Rabbit
Immunogen A synthesized peptide derived from human RAB15(Accession P59190), corresponding to amino acid residues K101-S151.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 104886, 299156, 376267
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.