Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Rab17 Antibody (OTI5E2), Alexa Fluor™ 647, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273765AF647
Description
Rab17 Monoclonal specifically detects Rab17 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
Rab17 | |
Monoclonal | |
Alexa Fluor 647 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
RAB17 | |
Full length recombinant protein of human Rab17 (NP_071894) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human | |
IgG1 |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
OTI5E2 | |
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
FLJ12538, RAB17, member RAS oncogene family, ras-related protein Rab-17 | |
Mouse | |
Affinity Purified | |
Signal Transduction | |
64284.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction