Learn More
RAB17 Mouse anti-Human, Clone: OTI3A5, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF501240

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
The Rab subfamily of small GTPases plays an important role in the regulation of membrane trafficking. RAB17 is an epithelial cell-specific GTPase.Specifications
RAB17 | |
Monoclonal | |
Unconjugated | |
RAB17 | |
RAB17, member RAS oncogene family | |
Mouse | |
Affinity Chromatography | |
RUO | |
64284 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
OTI3A5 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q9H0T7 | |
RAB17 | |
Full length human recombit protein of human RAB17 produced in HEK293T cell. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.