Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RAB17 Polyclonal Antibody

Goat Polyclonal Antibody
Supplier: Invitrogen™ PA518876
Description
This antibody is predicted to react with rat based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 64,000.
The Rab subfamily of small GTPases plays an important role in the regulation of membrane trafficking. RAB17 is an epithelial cell-specific GTPase.
Specifications
RAB17 | |
Polyclonal | |
Unconjugated | |
Rab17 | |
AW413472; Rab17; RAB17, member RAS oncogene family; ras-related protein Rab-17; RGD1562478 | |
Goat | |
Ammonium sulfate precipitation | |
RUO | |
19329 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
P35292 | |
Rab17 | |
Synthetic peptide sequence (RKDSFHKAQQWLED) corresponding to the internal amino acids of Rab17 (aa 102-116). | |
100 μg | |
Primary | |
Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction