Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RAB17 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA576280
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
The Rab subfamily of small GTPases plays an important role in the regulation of membrane trafficking. RAB17 is an epithelial cell-specific GTPase.
Specifications
RAB17 | |
Polyclonal | |
Unconjugated | |
Rab17 | |
AW413472; Rab17; RAB17, member RAS oncogene family; ras-related protein Rab-17; RGD1562478 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
19329, 503269, 64284 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
P35292, Q9H0T7 | |
Rab17 | |
Recombinant protein of human RAB17. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction