Learn More
Invitrogen™ RAB18 Monoclonal Antibody (1573CT811.119.39)

Description
RAB18 Monoclonal Antibody for Western Blot, Flow
Specifications
Specifications
| Antigen | RAB18 |
| Applications | Flow Cytometry, Western Blot |
| Classification | Monoclonal |
| Clone | 1573CT811.119.39 |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | Rab18 |
| Gene Accession No. | P35293, Q5EB77, Q9NP72 |
| Gene Alias | AA959686; GTP-binding protein Rab18; RAB18; RAB18 small GTPase; RAB18, member RAS oncogene family; RAB18LI1; Ras-related protein Rab-18; ras-related protein rab-18-like protein; WARBM3 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.