Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RAB18 Monoclonal Antibody (OTI7C9), OriGene
SDP

Catalog No. 502543895
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-3895 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-254-3895 Supplier OriGene Supplier No. CF811906
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

RAB18 Monoclonal antibody specifically detects RAB18 in Human samples. It is validated for Western Blot

Specifications

Antigen RAB18
Applications Western Blot
Classification Monoclonal
Clone OTI7C9
Concentration See Label
Conjugate Unconjugated
Formulation PBS with 8% trehalose and no preservative; pH 7.3
Gene ACSL5
Gene Accession No. Q9ULC5
Gene Alias AA959686; GTP-binding protein Rab18; RAB18; RAB18 small GTPase; RAB18, member RAS oncogene family; RAB18LI1; Ras-related protein Rab-18; ras-related protein rab-18-like protein; WARBM3
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 104-620 of human ACSL5 (NP_976313) produced in E.coli.
Purification Method Protein A/G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51703
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Lyophilized
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.