Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RAB18 Mouse anti-Human, Clone: 3A8, Bioss
SDP

Catalog No. 501979849
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-197-9849 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-197-9849 Supplier Bioss Supplier No. BSM51333M
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The protein encoded by this gene is a member of a family of Ras-related small GTPases that regulate membrane trafficking in organelles and transport vesicles. Knockdown studies is zebrafish suggest that this protein may have a role in eye and brain development. Mutations in this gene are associated with Warburg micro syndrome type 3. Alternatively spliced transcript variants have been found for this gene.

Specifications

Antigen RAB18
Applications Flow Cytometry, Western Blot
Classification Monoclonal
Clone 3A8
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation TBS with 1% BSA, 40% glycerol and 0.05% sodium azide; pH
Gene Rab18
Gene Accession No. Q9NP72
Gene Alias AA959686; BOS_12933; GTP-binding protein Rab18; LOC100356575; Rab18; RAB18 small GTPase; RAB18, member RAS oncogene family; RAB18LI1; Ras-related protein Rab-18; WARBM3
Gene Symbols Rab18
Host Species Mouse
Immunogen This RAB18 antibody is generated from a mouse immunized with a recombinant protein of human RAB18.
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22931
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.