Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RAB18 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA540654
Description
Peptide sequence: LFIEASAKTC DGVQCAFEEL VEKIIQTPGL WESENQNKGV KLSHREEGQG Sequence homology: Cow: 92%; Dog: 92%; Goat: 92%; Guinea Pig: 92%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 92%; Rat: 92%.
The protein encoded by this gene is a member of a family of Ras-related small GTPases that regulate membrane trafficking in organelles and transport vesicles. Knockdown studies is zebrafish suggest that this protein may have a role in eye and brain development. Mutations in this gene are associated with Warburg micro syndrome type 3. Alternatively spliced transcript variants have been found for this gene.
Specifications
RAB18 | |
Polyclonal | |
Unconjugated | |
Rab18 | |
AA959686; GTP-binding protein Rab18; RAB18; RAB18 small GTPase; RAB18, member RAS oncogene family; RAB18LI1; Ras-related protein Rab-18; ras-related protein rab-18-like protein; WARBM3 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
22931 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9NP72 | |
Rab18 | |
Synthetic peptide directed towards the C-terminal of human RAB18 (aa 146-195). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction