Learn More
Invitrogen™ RAB18 Polyclonal Antibody

Description
Immunogen sequence: VYDVTRRDTF VKLDNWLNEL ETYCTRNDIV NMLVGNKIDK ENREVDRNEG LKFARKHSML FIEASAKTCD GVQCAFEELV EKIIQTPGLW ESENQNKGVK LSHREEGQGG GACGGYCSV Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 97%.
Specifications
Specifications
| Antigen | RAB18 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.05 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | Rab18 |
| Gene Accession No. | Q9NP72 |
| Gene Alias | AA959686; GTP-binding protein Rab18; RAB18; RAB18 small GTPase; RAB18, member RAS oncogene family; RAB18LI1; Ras-related protein Rab-18; ras-related protein rab-18-like protein; WARBM3 |
| Gene Symbols | Rab18 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.