Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RAB19 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA562310
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562310 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562310 Supplier Invitrogen™ Supplier No. PA562310
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LIGNKCDLWE KRHVLFEDAC TLAEKYGLLA VLETSAKES Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 97%.

RAB19 gene ontology annotations related to this gene include endoplasmic reticulum to Golgi vesicle-mediated transport; intracellular protein transport; Rab protein signal transduction.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RAB19
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RAB19
Gene Accession No. A4D1S5
Gene Alias GTP-binding protein RAB19B; hypothetical protein LOC506911; Rab19; RAB19, member RAS oncogene family; RAB19B; Ras-related protein Rab-19; ras-related protein Rab-19-like; zmp:0000001111
Gene Symbols RAB19
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human RAB19. Recombinant protein control fragment (Product #RP-100960).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 401409
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.