Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RAB1A Monoclonal Antibody (4G10), PerCP
GREENER_CHOICE

Catalog No. PIMA545968
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA545968 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA545968 Supplier Invitrogen™ Supplier No. MA545968
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

A 1:1,000 dilution of MA5-45968 was sufficient for detection of RAB1A in 10 μg (A549, MEF); 20 μg (Mouse Brain) lysates by ECL immunoblot analysis. Detects approximately 23 kDa. Cross-reacts with RAB1B and RAB1C.

Ribosomes, the organelles that catalyze protein synthesis, consist of a small 40S subunit and a large 60S subunit. Together these subunits are composed of 4 RNA species and approximately 80 structurally distinct proteins. This gene encodes a member of the L18AE family of ribosomal proteins that is a component of the 60S subunit. The encoded protein may play a role in viral replication by interacting with the hepatitis C virus internal ribosome entry site (IRES). This gene is co-transcribed with the U68 snoRNA, located within the third intron. As is typical for genes encoding ribosomal proteins, there are multiple processed pseudogenes of this gene dispersed throughout the genome.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RAB1A
Applications Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 4G10
Concentration 1 mg/mL
Conjugate PerCP
Formulation 2.48mM MES, 95.64mM phosphate with 0.5M EDTA and no preservative; pH 7.4
Gene RAB1A
Gene Accession No. P62820, P62821
Gene Alias Ac2-048; DKFZP564B163; Gtbp; GTP binding protein Rab1a; LOW QUALITY PROTEIN: ras-related protein Rab-1A; mKIAA3012; Rab GTPase YPT1 homolog; RAB1; Rab-1; RAB1, member RAS oncogene family; Rab1A; RAB1A, member RAS oncogene family; RAB1A-like protein; Rab1r; Ras-related protein Rab-1A; ras-related YPT1 protein; YPT1; YPT1-related protein
Gene Symbols RAB1A
Host Species Mouse
Immunogen Full length recombinant RAB1A protein.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 19324, 5861
Target Species Human, Mouse
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.