Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RAB1A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5104066
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5104066 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5104066 Supplier Invitrogen™ Supplier No. PA5104066
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total RAB1A.

Ribosomes, the organelles that catalyze protein synthesis, consist of a small 40S subunit and a large 60S subunit. Together these subunits are composed of 4 RNA species and approximately 80 structurally distinct proteins. This gene encodes a member of the L18AE family of ribosomal proteins that is a component of the 60S subunit. The encoded protein may play a role in viral replication by interacting with the hepatitis C virus internal ribosome entry site (IRES). This gene is co-transcribed with the U68 snoRNA, located within the third intron. As is typical for genes encoding ribosomal proteins, there are multiple processed pseudogenes of this gene dispersed throughout the genome.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RAB1A
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene RAB1A
Gene Accession No. P62820, P62821, Q6NYB7
Gene Alias Ac2-048; DKFZP564B163; Gtbp; GTP binding protein Rab1a; LOW QUALITY PROTEIN: ras-related protein Rab-1A; mKIAA3012; Rab GTPase YPT1 homolog; RAB1; Rab-1; RAB1, member RAS oncogene family; Rab1A; RAB1A, member RAS oncogene family; RAB1A-like protein; Rab1r; Ras-related protein Rab-1A; ras-related YPT1 protein; YPT1; YPT1-related protein
Gene Symbols RAB1A
Host Species Rabbit
Immunogen A synthesized peptide derived from human RAB1A(Accession P62820), corresponding to amino acid residues Q163-C205.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 19324, 5861, 81754
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.