Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RAB1B Polyclonal Antibody
RAB1B Polyclonal Antibody
Supplier: Thermo Scientific PA596708
Description
RAB1B Polyclonal Antibody for Western Blot, IHC (P)

Specifications
RAB1B | |
Polyclonal | |
Unconjugated | |
RAB1B | |
1110011F09Rik; RAB1B; RAB1B, member RAS oncogene family; Rab1bl; RAB1B-like, member RAS oncogene family; ras-related protein Rab-1B; small GTP-binding protein | |
Rabbit | |
Affinity Chromatography | |
RUO | |
100126191, 76308, 81876 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
0.44 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P10536, Q9D1G1, Q9H0U4 | |
RAB1B | |
Recombinant fusion protein containing a sequence corresponding to amino acids 7-201 of human RAB1B (NP_1122431). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction