Learn More
RAB2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 154201AP150UL

Description
This antibody can recognize both RAB2A and RAB2B.
Members of the Rab protein family are nontransforming monomeric GTP-binding proteins of the Ras superfamily that contain 4 highly conserved regions involved in GTP binding and hydrolysis. Rabs are prenylated, membrane-bound proteins involved in vesicular fusion and trafficking. The mammalian RAB proteins show striking similarities to the S. cerevisiae YPT1 and SEC4 proteins, Ras-related GTP-binding proteins involved in the regulation of secretion.Specifications
RAB2 | |
Polyclonal | |
Unconjugated | |
RAB2A | |
RAB2, RAB2A, Ras related protein Rab 2A | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
305853, 5862, 59021, 65158, 76338, 84932 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.14 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P05712, P53994, P59279, P61019, Q8WUD1 | |
Rab2a, RAB2B | |
RAB2 Fusion Protein Ag7708 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.